2-Methoxy-5-(pentafluorthio)anilin 98%
Lieferant: Apollo Scientific
PC302255-1G
APOSPC302255-1GEA
0
EUR
APOSPC302255-1G
2-Methoxy-5-(pentafluorthio)anilin 98%
2-Methoxy-5-(pentafluorthio)anilin
Formel:
C₇H₈F₅NOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652485 CAS-Nummer: 1211587-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...