2-Methoxy-5-(trifluoromethoxy)benzamide 97%

Lieferant: Apollo Scientific
Achtung

PC302256-1G
APOSPC302256-1GEA 65.4 EUR
APOSPC302256-1G
2-Methoxy-5-(trifluoromethoxy)benzamide 97%
2-Methoxy-5-(trifluoromethoxy)benzamide
Formel: C₉H₈F₃NO₃
Molecular Weight: 235,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115962
CAS-Nummer: 886499-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR