2-Methoxy-6-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific

PC302260-1G
APOSPC302260-1GEA 377 EUR
APOSPC302260-1G
2-Methoxy-6-(trifluormethyl)benzylamin 98%
2-Methoxy-6-(trifluormethyl)benzylamin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832323
CAS-Nummer: 1017778-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR