3-(Difluormethoxy)zimtsäure 97%
Lieferant: Apollo Scientific
PC302267-1G
APOSPC302267-1GEA
0
EUR
APOSPC302267-1G
3-(Difluormethoxy)zimtsäure 97%
3-(Difluormethoxy)zimtsäure
Formel:
C₁₀H₈F₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06205319 CAS-Nummer: 630424-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...