3,5-Difluoro-2-methoxycinnamic acid 97%
Lieferant: Apollo Scientific
PC302270-1G
APOSPC302270-1GEA
102
EUR
APOSPC302270-1G
3,5-Difluoro-2-methoxycinnamic acid 97%
3,5-Difluoro-2-methoxycinnamic acid
Formel:
C₁₀H₈F₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999480 CAS-Nummer: 1092460-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...