3,5-Difluor-o-toluylsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Difluoro-2-methylbenzoic acid

PC302272-1G PC302272-5G PC302272-10G
APOSPC302272-1GEA 128 EUR
APOSPC302272-1G APOSPC302272-5G APOSPC302272-10G
3,5-Difluor-o-toluylsäure 98%
3,5-Difluor-o-toluylsäure
Formel: C₈H₆F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832334
CAS-Nummer: 1003710-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR