3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC302285-1G PC302285-5G PC302285-10G PC302285-25G
APOSPC302285-1GEA 231 EUR
APOSPC302285-1G APOSPC302285-5G APOSPC302285-10G APOSPC302285-25G
3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl bromide 97%
3-Chloro-2-fluoro-4-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrClF₄
Lagertemperatur: Raumtemperatur
MDL: MFCD23795329
CAS-Nummer: 1431329-66-6
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR