3-Chloro-5-fluoro-4-methoxyaniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302292-1G
APOSPC302292-1GEA
120
EUR
APOSPC302292-1G
3-Chloro-5-fluoro-4-methoxyaniline 97%
3-Chloro-5-fluoro-4-methoxyaniline
Formel:
C₇H₇ClFNO Molecular Weight: 175,588 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832236 CAS-Nummer: 62918-78-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...