3'-Fluor-2'-(trifluormethyl)acetophenon 98%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Fluoro-2-(trifluoromethyl)phenyl)ethanone

PC302298-1G PC302298-5G PC302298-10G PC302298-25G
APOSPC302298-1GEA 154 EUR
APOSPC302298-1G APOSPC302298-5G APOSPC302298-10G APOSPC302298-25G
3'-Fluor-2'-(trifluormethyl)acetophenon 98%
3'-Fluor-2'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD09832249
CAS-Nummer: 1017777-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR