3'-Fluor-5'-(trifluormethoxy)acetophenon 97%

Lieferant: Apollo Scientific
Achtung

PC302302-1G PC302302-5G
APOSPC302302-1GEA 0 EUR
APOSPC302302-1G APOSPC302302-5G
3'-Fluor-5'-(trifluormethoxy)acetophenon 97%
3'-Fluor-5'-(trifluormethoxy)acetophenon
Formel: C₉H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18783230
CAS-Nummer: 1352999-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR