3-Fluor-5-(trifluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC302304-1G
APOSPC302304-1GEA 0 EUR
APOSPC302304-1G
3-Fluor-5-(trifluormethoxy)benzylalkohol 98%
3-Fluor-5-(trifluormethoxy)benzylalkohol
Formel: C₈H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18783225
CAS-Nummer: 86256-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR