2-Fluoro-5-methyl-4-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC302311-1G
APOSPC302311-1GEA 0 EUR
APOSPC302311-1G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzenesulphonyl chloride
2-Fluoro-5-methyl-4-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₈H₅ClF₄O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28054244
CAS-Nummer: 1706431-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR