3-Methyl-5-(trifluormethyl)benzylamin 97%

Lieferant: Apollo Scientific

PC302312-1G
APOSPC302312-1GEA 0 EUR
APOSPC302312-1G
3-Methyl-5-(trifluormethyl)benzylamin 97%
3-Methyl-5-(trifluormethyl)benzylamin
Formel: C₉H₁₀F₃N
Molecular Weight: 189,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213553
CAS-Nummer: 292151-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR