4-(Difluormethoxy)-2-fluorbenzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC302314-1G
APOSPC302314-1GEA 313 EUR
APOSPC302314-1G
4-(Difluormethoxy)-2-fluorbenzonitril 97%
4-(Difluormethoxy)-2-fluorbenzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832366
CAS-Nummer: 1017779-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR