4-(Difluoromethoxy)-3-methoxycinnamic acid 97%

Lieferant: Apollo Scientific

PC302315-1G
APOSPC302315-1GEA 92.8 EUR
APOSPC302315-1G
4-(Difluoromethoxy)-3-methoxycinnamic acid 97%
4-(Difluoromethoxy)-3-methoxycinnamic acid
Formel: C₁₁H₁₀F₂O₄
Molecular Weight: 244,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01794575
CAS-Nummer: 324579-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR