4-(Difluormethoxy)zimtsäure 97%

Lieferant: Apollo Scientific

PC302316-1G PC302316-5G
APOSPC302316-1GEA 0 EUR
APOSPC302316-1G APOSPC302316-5G
4-(Difluormethoxy)zimtsäure 97%
4-(Difluormethoxy)zimtsäure
Formel: C₁₀H₈F₂O₃
Schmelzpunkt: 183…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01222822
CAS-Nummer: 103962-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR