4-Chlor-2-(trifluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC302320-1G PC302320-5G
APOSPC302320-1GEA 92.8 EUR
APOSPC302320-1G APOSPC302320-5G
4-Chlor-2-(trifluormethoxy)benzylalkohol 98%
4-Chlor-2-(trifluormethoxy)benzylalkohol
Formel: C₈H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18393819
CAS-Nummer: 1261673-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR