4-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302323-1G PC302323-5G
APOSPC302323-1GEA 0 EUR
APOSPC302323-1G APOSPC302323-5G
4-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%
4-Chloro-2-fluoro-3-methoxybenzoyl chloride
Formel: C₈H₅Cl₂FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19687205
CAS-Nummer: 1323966-24-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR