4-Chloro-3-ethoxy-2-fluorobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-3-ethoxy-2-fluorobenzyl alcohol

PC302328-1G
APOSPC302328-1GEA 93.1 EUR
APOSPC302328-1G
4-Chloro-3-ethoxy-2-fluorobenzyl alcohol 97%
4-Chloro-3-ethoxy-2-fluorobenzyl alcohol
Formel: C₉H₁₁ClFO₂
Molecular Weight: 204.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687185
CAS-Nummer: 1323955-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR