4-Fluor-2-(trifluormethoxy)benzylalkohol

Lieferant: Apollo Scientific

PC302339-1G PC302339-5G
APOSPC302339-1GEA 180 EUR
APOSPC302339-1G APOSPC302339-5G
4-Fluor-2-(trifluormethoxy)benzylalkohol
4-Fluor-2-(trifluormethoxy)benzylalkohol
Formel: C₈H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19687174
CAS-Nummer: 1242252-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR