4-Fluor-2-(trifluormethoxy)benzylamin 98%

Lieferant: Apollo Scientific

PC302340-1G PC302340-5G PC302340-25G
APOSPC302340-1GEA 119 EUR
APOSPC302340-1G APOSPC302340-5G APOSPC302340-25G
4-Fluor-2-(trifluormethoxy)benzylamin 98%
4-Fluor-2-(trifluormethoxy)benzylamin
Formel: C₈H₇F₄NO
Molecular Weight: 209.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687190
CAS-Nummer: 1241828-99-8
UN: 3267
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR