4-Methoxy-3-(trifluormethoxy)benzonitril 97%
Lieferant: Apollo Scientific
PC302344-1G
APOSPC302344-1GEA
132
EUR
APOSPC302344-1G
4-Methoxy-3-(trifluormethoxy)benzonitril 97%
4-Methoxy-3-(trifluormethoxy)benzonitril
Formel:
C₉H₆F₃NO₂ Molecular Weight: 217,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398458 CAS-Nummer: 1261653-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...