4-Methoxy-3-(trifluormethoxy)benzylalkohol

Lieferant: Apollo Scientific

PC302346-1G PC302346-5G
APOSPC302346-1GEA 0 EUR
APOSPC302346-1G APOSPC302346-5G
4-Methoxy-3-(trifluormethoxy)benzylalkohol
4-Methoxy-3-(trifluormethoxy)benzylalkohol
Formel: C₉H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18398472
CAS-Nummer: 853771-91-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR