4-Methoxy-3-(trifluoromethoxy)cinnamic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-3-(trifluoromethoxy)cinnamic acid

PC302347-1G
APOSPC302347-1GEA 168 EUR
APOSPC302347-1G
4-Methoxy-3-(trifluoromethoxy)cinnamic acid 97%
4-Methoxy-3-(trifluoromethoxy)cinnamic acid
Formel: C₁₁H₉F₃O₄
Molecular Weight: 262,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398503
CAS-Nummer: 1262018-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR