2-Fluoro-4-(trifluoromethoxy)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC302352-1G
APOSPC302352-1GEA 1740 EUR
APOSPC302352-1G
2-Fluoro-4-(trifluoromethoxy)benzenesulphonyl chloride
2-Fluoro-4-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₇H₃ClF₄O₃S
Molecular Weight: 278,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054238
CAS-Nummer: 954388-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR