5-Chlor-2-(trifluormethoxy)benzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
PC302354-1G
PC302354-5G
APOSPC302354-1GEA
157
EUR
APOSPC302354-1G
APOSPC302354-5G
5-Chlor-2-(trifluormethoxy)benzylalkohol
5-Chlor-2-(trifluormethoxy)benzylalkohol
Formel:
C₈H₆ClF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519357 CAS-Nummer: 874821-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...