5-Chloro-2-(trifluoromethoxy)cinnamic acid

Lieferant: Apollo Scientific
Achtung

PC302355-1G
APOSPC302355-1GEA 0 EUR
APOSPC302355-1G
5-Chloro-2-(trifluoromethoxy)cinnamic acid
5-Chloro-2-(trifluoromethoxy)cinnamic acid
Formel: C₁₀H₆ClF₃O₃
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11519361
CAS-Nummer: 1092461-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR