5-Fluor-2-(trifluormethoxy)benzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
PC302358-1G
PC302358-5G
APOSPC302358-1GEA
102
EUR
APOSPC302358-1G
APOSPC302358-5G
5-Fluor-2-(trifluormethoxy)benzylalkohol
5-Fluor-2-(trifluormethoxy)benzylalkohol
Formel:
C₈H₆F₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519336 CAS-Nummer: 1092460-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...