5-Fluor-2-(trifluormethoxy)benzylalkohol

Lieferant: Apollo Scientific
Achtung

PC302358-1G PC302358-5G
APOSPC302358-1GEA 102 EUR
APOSPC302358-1G APOSPC302358-5G
5-Fluor-2-(trifluormethoxy)benzylalkohol
5-Fluor-2-(trifluormethoxy)benzylalkohol
Formel: C₈H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11519336
CAS-Nummer: 1092460-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR