5-Fluoro-2-(trifluoromethoxy)cinnamic acid 97%
Lieferant: Apollo Scientific
PC302359-1G
PC302359-5G
PC302359-250MG
APOSPC302359-1GEA
386
EUR
APOSPC302359-1G
APOSPC302359-5G
APOSPC302359-250MG
5-Fluoro-2-(trifluoromethoxy)cinnamic acid 97%
5-Fluoro-2-(trifluoromethoxy)cinnamic acid
Formel:
C₁₀H₆F₄O₃ Molecular Weight: 250.146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519340 CAS-Nummer: 1092460-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...