5-Methyl-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific

PC302362-1G PC302362-5G
APOSPC302362-1GEA 92.8 EUR
APOSPC302362-1G APOSPC302362-5G
5-Methyl-2-(trifluormethyl)benzylbromid 98%
5-Methyl-2-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660296
CAS-Nummer: 886502-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR