6-Chloro-2,3-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2,3-difluorophenylacetonitrile

PC302366-1G
APOSPC302366-1GEA 87 EUR
APOSPC302366-1G
6-Chloro-2,3-difluorophenylacetonitrile 97%
6-Chloro-2,3-difluorophenylacetonitrile
Formel: C₈H₄ClF₂N
Molecular Weight: 187,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201041
CAS-Nummer: 1373920-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR