6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde

PC302370-1G PC302370-5G PC302370-25G
APOSPC302370-1GEA 87 EUR
APOSPC302370-1G APOSPC302370-5G APOSPC302370-25G
6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde 98%
6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
Formel: C₉H₄F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD22201035
CAS-Nummer: 1373920-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR