(2,6-Difluor-3-methoxyphenyl)essigsäure 97%

Lieferant: Apollo Scientific

PC302380-1G
APOSPC302380-1GEA 248 EUR
APOSPC302380-1G
(2,6-Difluor-3-methoxyphenyl)essigsäure 97%
(2,6-Difluor-3-methoxyphenyl)essigsäure
Formel: (C₆H₂F₂OCH₃)CH₂COOH
Molecular Weight: 186,16 g/mol
Schmelzpunkt: 124…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115908
CAS-Nummer: 886498-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR