2-Methoxy-4-(trifluoromethyl)benzamide 98%

Lieferant: Apollo Scientific

PC302389-1G
APOSPC302389-1GEA 130 EUR
APOSPC302389-1G
2-Methoxy-4-(trifluoromethyl)benzamide 98%
2-Methoxy-4-(trifluoromethyl)benzamide
Formel: C₉H₈F₃NO₂
Molecular Weight: 219,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115978
CAS-Nummer: 886500-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR