3-Chloro-5-(trifluoromethyl)benzoyl chloride 98%

Lieferant: Apollo Scientific
Gefahr

PC302398-5G
APOSPC302398-5GEA 136 EUR
APOSPC302398-5G
3-Chloro-5-(trifluoromethyl)benzoyl chloride 98%
3-Chloro-5-(trifluoromethyl)benzoyl chloride
Formel: C₈H₃Cl₂F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04115847
CAS-Nummer: 886496-83-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR