5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
PC302409-1G
PC302409-250MG
APOSPC302409-1GEA
0
EUR
APOSPC302409-1G
APOSPC302409-250MG
5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulphonyl chloride 97%
5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulphonyl chloride
Formel:
C₈H₅Cl₂F₃O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28054251 CAS-Nummer: 1706447-06-4 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...