4-Chloro-3-(trifluoromethyl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302411-5G PC302411-1G
APOSPC302411-5GEA 0 EUR
APOSPC302411-5G APOSPC302411-1G
4-Chloro-3-(trifluoromethyl)benzoyl chloride 97%
4-Chloro-3-(trifluoromethyl)benzoyl chloride
Formel: C₈H₃Cl₂F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04115833
CAS-Nummer: 1735-55-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR