4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
PC302414-1G
PC302414-250MG
PC302414-5G
APOSPC302414-1GEA
237
EUR
APOSPC302414-1G
APOSPC302414-250MG
APOSPC302414-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid
Formel:
C₁₀H₈F₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201074 CAS-Nummer: 1373920-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...