6-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302416-25G PC302416-5G PC302416-1G
APOSPC302416-25GEA 0 EUR
APOSPC302416-25G APOSPC302416-5G APOSPC302416-1G
6-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%
6-Chloro-2-fluoro-3-methoxybenzoyl chloride
Formel: C₈H₅Cl₂FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115948
CAS-Nummer: 886499-63-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR