2-(Difluormethoxy)-4-fluorbenzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302419-1G PC302419-5G PC302419-10G
APOSPC302419-1GEA 128 EUR
APOSPC302419-1G APOSPC302419-5G APOSPC302419-10G
2-(Difluormethoxy)-4-fluorbenzylbromid 98%
2-(Difluormethoxy)-4-fluorbenzylbromid
Formel: C₈H₆BrF₃O
Molecular Weight: 255.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832363
CAS-Nummer: 1017779-42-8
UN: 3261
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR