2-(Trifluoromethoxy)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302422-1G PC302422-5G PC302422-250MG
APOSPC302422-1GEA 0 EUR
APOSPC302422-1G APOSPC302422-5G APOSPC302422-250MG
2-(Trifluoromethoxy)-DL-phenylalanine
2-(Trifluoromethoxy)-DL-phenylalanine
Formel: C₁₀H₁₀F₃NO₃
Schmelzpunkt: 236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07371965
CAS-Nummer: 1218228-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR