2,3,5-Trifluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302425-1G PC302425-5G PC302425-25G PC302425-500MG
APOSPC302425-1GEA 128 EUR
APOSPC302425-1G APOSPC302425-5G APOSPC302425-25G APOSPC302425-500MG
2,3,5-Trifluoro-DL-phenylalanine
2,3,5-Trifluoro-DL-phenylalanine
Formel: C₉H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116036
CAS-Nummer: 1259960-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR