2,3,5-Trifluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302425-1G
PC302425-5G
PC302425-25G
PC302425-500MG
APOSPC302425-1GEA
128
EUR
APOSPC302425-1G
APOSPC302425-5G
APOSPC302425-25G
APOSPC302425-500MG
2,3,5-Trifluoro-DL-phenylalanine
2,3,5-Trifluoro-DL-phenylalanine
Formel:
C₉H₈F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116036 CAS-Nummer: 1259960-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...