2,4-Difluor-3-methoxybenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302438-25G
PC302438-5G
PC302438-1G
APOSPC302438-25GEA
366
EUR
APOSPC302438-25G
APOSPC302438-5G
APOSPC302438-1G
2,4-Difluor-3-methoxybenzoesäure 98%
2,4-Difluoro-3-methoxybenzoic acid
Formel:
C₈H₆F₂O₃ Schmelzpunkt: 195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258905 CAS-Nummer: 178974-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...