2-Chlor-6-(trifluormethoxy)benzonitril

Lieferant: Apollo Scientific
Achtung

PC302444-1G
APOSPC302444-1GEA 0 EUR
APOSPC302444-1G
2-Chlor-6-(trifluormethoxy)benzonitril
2-Chlor-6-(trifluormethoxy)benzonitril
Formel: C₈H₃ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18393805
CAS-Nummer: 1261779-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR