2-Fluor-4-(pentafluorthio)benzamid 97%

Lieferant: Apollo Scientific

PC302451-1G
APOSPC302451-1GEA 348 EUR
APOSPC302451-1G
2-Fluor-4-(pentafluorthio)benzamid 97%
2-Fluor-4-(pentafluorthio)benzamid
Formel: C₇H₅F₆NOS
Molecular Weight: 265,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652504
CAS-Nummer: 1240257-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR