2-Fluor-4-(trifluormethoxy)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302454-1G
PC302454-5G
PC302454-10G
APOSPC302454-1GEA
0
EUR
APOSPC302454-1G
APOSPC302454-5G
APOSPC302454-10G
2-Fluor-4-(trifluormethoxy)benzylbromid 98%
2-Fluor-4-(trifluormethoxy)benzylbromid
Formel:
C₈H₅BrF₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652442 CAS-Nummer: 1240257-47-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...