2-Fluor-4-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302454-1G PC302454-5G PC302454-10G
APOSPC302454-1GEA 0 EUR
APOSPC302454-1G APOSPC302454-5G APOSPC302454-10G
2-Fluor-4-(trifluormethoxy)benzylbromid 98%
2-Fluor-4-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD16652442
CAS-Nummer: 1240257-47-9
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR