2-Fluoro-4-methyl-DL-phenylglycine

Lieferant: Apollo Scientific

PC302456-1G
APOSPC302456-1GEA 284 EUR
APOSPC302456-1G
2-Fluoro-4-methyl-DL-phenylglycine
2-Fluoro-4-methyl-DL-phenylglycine
Formel: C₉H₁₀FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116045
CAS-Nummer: 1039118-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR